(11aR)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%

Supplier: Ambeed
Warning

A1006112-100MG A1006112-1G A1006112-250MG
76646-568EA 84 USD
76646-568 76646-570 76646-572
(11aR)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%
(11aR)-N,N-Diisopropyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine

Precaución: for Research Use Only

Formula: C₂₃H₂₈NO₂P
MW: 381.45 g/mol
CAS Number: 500997-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR