(11aR)-N,N-Dicyclohexyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%

Supplier: Ambeed
Warning

A1006111-100MG A1006111-1G A1006111-250MG
76646-562EA 46 USD
76646-562 76646-564 76646-566
(11aR)-N,N-Dicyclohexyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine 97%
(11aR)-N,N-Dicyclohexyl-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-amine

Precaución: for Research Use Only

Formula: C₂₉H₃₆NO₂P
MW: 461.58 g/mol
CAS Number: 500997-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR