(11bR)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%

Supplier: Ambeed
Warning

A1006099-100MG A1006099-1G A1006099-250MG
76646-552EA 63 USD
76646-552 76646-554 76646-556
(11bR)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 97%
(11bR)-4-Hydroxy-2,6-bis(4-nitrophenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₃₂H₂₇N₂O₈P
MW: 598.5392 g/mol
MDL Number: MFCD31747031
CAS Number: 791616-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR