(11bS)-1,1,1-Trifluoro-N-(4-oxido-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 97%

Supplier: Ambeed
Warning

A1006061-100MG A1006061-1G A1006061-250MG A1006061-50MG
76646-492EA 204 USD
76646-492 76646-494 76646-496 76646-498
(11bS)-1,1,1-Trifluoro-N-(4-oxido-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 97%
(11bS)-1,1,1-Trifluoro-N-(4-oxido-2,6-di(phenanthren-9-yl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₄₉H₂₉F₃NO₅PS
CAS Number: 1256334-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR