((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(di(naphthalen-1-yl)methanol) ≥95%, ee 99%

Supplier: Ambeed
Warning

A1006006-1G A1006006-250MG A1006006-25G A1006006-5G
76646-484EA 40 USD
76646-484 76646-486 76646-488 76646-490
((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(di(naphthalen-1-yl)methanol) ≥95%, ee 99%
((4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(di(naphthalen-1-yl)methanol)

Precaución: for Research Use Only

Formula: C₄₇H₃₈O₄
CAS Number: 171086-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR