2,6-Bis((R)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A1005968-100MG
A1005968-1G
A1005968-250MG
A1005968-5G
A1005968-25G
76646-478EA
12
USD
76646-478
76646-480
76646-482
77619-594
77782-801
2,6-Bis((R)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine ≥97%, ee 99%
2,6-Bis((R)-4-methyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₃H₁₅N₃O₂ |
CAS Number:
256377-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...