2,6-Bis((4R,5R)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A1005961-100MG A1005961-1G A1005961-250MG
76646-466EA 84.88 USD
76646-466 76646-468 76646-470
2,6-Bis((4R,5R)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl)pyridine 97%
2,6-Bis((4R,5R)-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₃₅H₂₉N₅
MW: 519.6383 g/mol
CAS Number: 863491-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR