(S,S)-iPr-BI-DIME 97%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-isopropyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

A1005933-100MG A1005933-250MG
76646-438EA 110.34 USD
76646-438 77619-580
(S,S)-iPr-BI-DIME 97%
(S,S)-iPr-BI-DIME

Precaución: for Research Use Only

Formula: C₂₂H₂₉O₃P
MW: 372.438 g/mol
MDL Number: MFCD32206571
CAS Number: 1477517-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR