(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide 97%

Supplier: Ambeed
Warning

A1005932-100MG A1005932-50MG A1005932-250MG
76646-434EA 141.1 USD
76646-434 76646-436 77595-672
(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide 97%
(R)-N-((S)-2-(Diphenylphosphanyl)-1-(2-(diphenylphosphanyl)phenyl)ethyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₆H₃₇NOP₂S
MW: 593.6976 g/mol
MDL Number: MFCD32671242
CAS Number: 1824731-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR