2,4,6-Tri(1H-imidazol-1-yl)-1,3,5-triazine 97%

Supplier: Ambeed
Warning

A1005903-100MG A1005903-250MG A1005903-5G
76646-428EA 41.38 USD
76646-428 76646-430 76646-432
2,4,6-Tri(1H-imidazol-1-yl)-1,3,5-triazine 97%
2,4,6-Tri(1H-imidazol-1-yl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₁₂H₉N₉
MW: 279.2602 g/mol
MDL Number: MFCD00412096
CAS Number: 14445-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR