7-Chlorodibenzo[c,h]acridine 97%

Supplier: Ambeed
Warning

A1005895-1G A1005895-250MG
76646-424EA 56.23 USD
76646-424 76646-426
7-Chlorodibenzo[c,h]acridine 97%
7-Chlorodibenzo[c,h]acridine

Precaución: for Research Use Only

Formula: C₂₁H₁₂ClN
MW: 313.7797 g/mol
MDL Number: MFCD32068124
CAS Number: 859745-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR