3-(2-(2-Bromoethoxy)ethoxy)prop-1-yne 97%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG2-bromide

A1005875-100MG A1005875-250MG A1005875-1G A1005875-5G A1005875-25G
76646-414EA 24 USD
76646-414 76646-416 77548-884 77570-052 77782-784
3-(2-(2-Bromoethoxy)ethoxy)prop-1-yne 97%
3-(2-(2-Bromoethoxy)ethoxy)prop-1-yne

Precaución: for Research Use Only

Formula: C₇H₁₁BrO₂
MDL Number: MFCD24465329
CAS Number: 1287660-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR