2-(2-(3-Aminopropoxy)ethoxy)ethan-1-ol 97%

Supplier: Ambeed
Danger

A1005548-100MG A1005548-1G A1005548-250MG
76646-396EA 23 USD
76646-396 76646-398 76646-400
2-(2-(3-Aminopropoxy)ethoxy)ethan-1-ol 97%
2-(2-(3-Aminopropoxy)ethoxy)ethan-1-ol

Precaución: for Research Use Only

Formula: C₇H₁₇NO₃
MDL Number: MFCD00661075
CAS Number: 112-33-4
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR