8-Methylquinoline 1-oxide 97%

Supplier: Ambeed
Warning

A1005442-100MG A1005442-250MG A1005442-1G
76646-392EA 46 USD
76646-392 76646-394 77619-570
8-Methylquinoline 1-oxide 97%
8-Methylquinoline 1-oxide

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD22666433
CAS Number: 4053-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR