4,4'-((4-Bromophenyl)azanediyl)dibenzaldehyde 97%

Supplier: Ambeed
Warning

A1005327-100MG A1005327-250MG A1005327-5G
76646-384EA 22.28 USD
76646-384 76646-386 76646-388
4,4'-((4-Bromophenyl)azanediyl)dibenzaldehyde 97%
4,4'-((4-Bromophenyl)azanediyl)dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₀H₁₄BrNO₂
MW: 380.2347 g/mol
MDL Number: MFCD31814477
CAS Number: 167859-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR