(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1005265-100MG A1005265-1G A1005265-250MG
76646-362EA 210.06 USD
76646-362 76646-364 76646-366
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
(11bR)-N-(2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₃₇H₁₇F₁₅NO₅PS
MW: 903.5494 g/mol
CAS Number: 1010800-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR