N-(2,6-Di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%

Supplier: Ambeed
Warning

A1005262-100MG A1005262-1G A1005262-250MG A1005262-50MG
76646-354EA 65 USD
76646-354 76646-356 76646-358 76646-360
N-(2,6-Di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 97%
N-(2,6-Di(naphthalen-1-yl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₄₁H₂₅F₃NO₅PS
CAS Number: 1226907-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR