4,5-Di(9H-carbazol-9-yl)phthalonitrile 97%

Supplier: Ambeed
Warning

A1005124-100MG A1005124-250MG A1005124-1G A1005124-50MG
76646-342EA 222 USD
76646-342 76646-344 77570-042 77595-664
4,5-Di(9H-carbazol-9-yl)phthalonitrile 97%
4,5-Di(9H-carbazol-9-yl)phthalonitrile

Precaución: for Research Use Only

Formula: C₃₂H₁₈N₄
MW: 458.51 g/mol
MDL Number: MFCD28369264
CAS Number: 1416881-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR