2,7-Di-tert-butyl-9H-carbazole 97%

Supplier: Ambeed
Warning

A1005104-1G A1005104-250MG A1005104-25G A1005104-5G A1005104-100G
76646-334EA 14 USD
76646-334 76646-336 77619-548 77619-550 77782-765
2,7-Di-tert-butyl-9H-carbazole 97%
2,7-Di-tert-butyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₀H₂₅N
MDL Number: MFCD32204944
CAS Number: 69386-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR