N,N-Bis(4'-diphenylamino-4-biphenylyl)amine 98%

Supplier: Ambeed
Warning

Synonyms: N'-[4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl]-N,N-diphenyl-[1,1'-biphenyl]-4,4'-diamine, N4-(4'-(Diphenylamino)-[1,1'-biphenyl]-4-yl)-N4',N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

A1005102-100MG A1005102-1G A1005102-250MG
76646-328EA 18 USD
76646-328 76646-330 76646-332
N,N-Bis(4'-diphenylamino-4-biphenylyl)amine 98%
N,N-Bis(4'-diphenylamino-4-biphenylyl)amine

Precaución: for Research Use Only

Formula: C₄₈H₃₇N₃
MW: 655.85 g/mol
MDL Number: MFCD32263460
CAS Number: 167218-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR