4,4'-(2-(4-Bromophenyl)-2-phenylethene-1,1-diyl)bis(methoxybenzene) 97%

Supplier: Ambeed
Warning

A1005000-1G A1005000-250MG A1005000-5G
76646-304EA 135.8 USD
76646-304 76646-306 76646-308
4,4'-(2-(4-Bromophenyl)-2-phenylethene-1,1-diyl)bis(methoxybenzene) 97%
4,4'-(2-(4-Bromophenyl)-2-phenylethene-1,1-diyl)bis(methoxybenzene)

Precaución: for Research Use Only

Formula: C₂₈H₂₃BrO₂
MW: 471.3850 g/mol
MDL Number: MFCD30187353
CAS Number: 1378502-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR