H-Lys(crotonyl)-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S,E)-2-Amino-6-(but-2-enamido)hexanoic acid

A1004962-100MG A1004962-1G A1004962-250MG A1004962-5G
76646-298EA 54.11 USD
76646-298 76646-300 76646-302 77595-658
H-Lys(crotonyl)-OH 97%
H-Lys(crotonyl)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
MDL Number: MFCD22381072
CAS Number: 1338823-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR