Benzyl 2,3,4-tri-O-benzyl-α-D-glucopyranoside 98%

Supplier: Ambeed
Warning

A1004952-1G A1004952-250MG A1004952-5G
76646-292EA 119.89 USD
76646-292 76646-294 76646-296
Benzyl 2,3,4-tri-O-benzyl-α-D-glucopyranoside 98%
Benzyl 2,3,4-tri-O-benzyl-α-D-glucopyranoside

Precaución: for Research Use Only

Formula: C₃₄H₃₆O₆
MW: 540.6461 g/mol
MDL Number: MFCD00061638
CAS Number: 59935-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR