3-(3-Aminopropoxy)propan-1-ol 97%

Supplier: Ambeed
Danger

A1004855-100MG A1004855-1G A1004855-250MG
76646-286EA 114.58 USD
76646-286 76646-288 76646-290
3-(3-Aminopropoxy)propan-1-ol 97%
3-(3-Aminopropoxy)propan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₅NO₂
MW: 133.1900 g/mol
MDL Number: MFCD12192584
CAS Number: 118273-16-8
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR