Cyclohexyl methanesulfonate 97%

Supplier: Ambeed
Warning

A1004834-100MG A1004834-1G A1004834-250MG A1004834-5G
76646-278EA 43.5 USD
76646-278 76646-280 76646-282 76646-284
Cyclohexyl methanesulfonate 97%
Cyclohexyl methanesulfonate

Precaución: for Research Use Only

Formula: C₇H₁₄O₃S
MW: 178.2493 g/mol
MDL Number: MFCD26961237
CAS Number: 16156-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR