Bis(2-methoxyphenyl)phosphine oxide 98%

Supplier: Ambeed
Warning

A1004831-1G A1004831-250MG A1004831-5G A1004831-25G
76646-272EA 64 USD
76646-272 76646-274 76646-276 77782-751
Bis(2-methoxyphenyl)phosphine oxide 98%
Bis(2-methoxyphenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₁₄H₁₅O₃P
MDL Number: MFCD33022345
CAS Number: 71360-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR