1-Decylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A1004782-1G A1004782-250MG
76646-264EA 52 USD
76646-264 76646-266
1-Decylboronic acid pinacol ester 98%
1-Decylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₃₃BO₂
MDL Number: MFCD12546186
CAS Number: 141091-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR