2-Biphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-([1,1'-Biphenyl]-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A1004780-1G A1004780-25G A1004780-5G
76646-258EA 27.59 USD
76646-258 76646-260 76646-262
2-Biphenylboronic acid pinacol ester 98%
2-Biphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₁BO₂
MW: 280.17 g/mol
MDL Number: MFCD14636459
CAS Number: 914675-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR