5-Bromo-2,3,4-trifluorobenzoic acid 97%

Supplier: Ambeed
Warning

A1003996-1G A1003996-25G A1003996-5G
76646-224EA 11.67 USD
76646-224 76646-226 76646-228
5-Bromo-2,3,4-trifluorobenzoic acid 97%
5-Bromo-2,3,4-trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂BrF₃O₂
MW: 254.989 g/mol
MDL Number: MFCD12756771
CAS Number: 212631-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR