Sodium 2,5-dimethylbenzenesulfonate 98%

Supplier: Ambeed
Warning

A1003843-1G A1003843-5G
76646-220EA 7 USD
76646-220 76646-222
Sodium 2,5-dimethylbenzenesulfonate 98%
Sodium 2,5-dimethylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₈H₉NaO₃S
MDL Number: MFCD00066377
CAS Number: 827-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR