4,4',4''-(1H-Imidazole-2,4,5-triyl)tripyridine 97%

Supplier: Ambeed
Warning

A1003742-100MG A1003742-250MG A1003742-5G
76646-208EA 19.1 USD
76646-208 76646-210 76646-212
4,4',4''-(1H-Imidazole-2,4,5-triyl)tripyridine 97%
4,4',4''-(1H-Imidazole-2,4,5-triyl)tripyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₃N₅
MW: 299.3293 g/mol
CAS Number: 23974-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR