1,4-Di(1H-imidazol-1-yl)benzene 97%

Supplier: Ambeed
Warning

A1003741-1G A1003741-250MG A1003741-5G
76646-202EA 57.29 USD
76646-202 76646-204 76646-206
1,4-Di(1H-imidazol-1-yl)benzene 97%
1,4-Di(1H-imidazol-1-yl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄
MW: 210.2346 g/mol
MDL Number: MFCD23103179
CAS Number: 25372-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR