1,3-Di(1H-imidazol-1-yl)propane 97%

Supplier: Ambeed
Warning

A1003739-250MG A1003739-1G
76646-200EA 20.16 USD
76646-200 77619-514
1,3-Di(1H-imidazol-1-yl)propane 97%
1,3-Di(1H-imidazol-1-yl)propane

Precaución: for Research Use Only

Formula: C₉H₁₂N₄
MW: 176.2184 g/mol
MDL Number: MFCD11110663
CAS Number: 69506-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR