(E)-5,5'-(Diazene-1,2-diyl)diisophthalic acid 97%

Supplier: Ambeed
Warning

A1003732-1G A1003732-250MG A1003732-5G A1003732-25G
76646-196EA 51.99 USD
76646-196 76646-198 77570-026 77619-512
(E)-5,5'-(Diazene-1,2-diyl)diisophthalic acid 97%
(E)-5,5'-(Diazene-1,2-diyl)diisophthalic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₀N₂O₈
MW: 358.26 g/mol
MDL Number: MFCD30738344
CAS Number: 1007597-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR