Sodium benzo[d]thiazole-2-sulfinate 97%

Supplier: Ambeed
Warning

A1003723-100MG A1003723-250MG
76646-192EA 146 USD
76646-192 76646-194
Sodium benzo[d]thiazole-2-sulfinate 97%
Sodium benzo[d]thiazole-2-sulfinate

Precaución: for Research Use Only

Formula: C₇H₄NNaO₂S₂
MW: 221.2319 g/mol
MDL Number: MFCD31442267
CAS Number: 61073-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR