1-(2-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A1324943-1G
A1324943-250MG
76657-874EA
308.73
USD
76657-874
76657-876
1-(2-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone 97%
1-(2-Bromoimidazo[1,2-a]pyridin-3-yl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₇BrN₂O MW: 239.0687 g/mol |
MDL Number:
MFCD22034752 CAS Number: 1367773-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...