4-Benzylidene-2,6-di-tert-butylcyclohexa-2,5-dienone 97%

Supplier: Ambeed
Warning

A1003591-100G A1003591-1G A1003591-5G
76646-174EA 402.09 USD
76646-174 76646-176 76646-178
4-Benzylidene-2,6-di-tert-butylcyclohexa-2,5-dienone 97%
4-Benzylidene-2,6-di-tert-butylcyclohexa-2,5-dienone

Precaución: for Research Use Only

Formula: C₂₁H₂₆O
MW: 294.4306 g/mol
MDL Number: MFCD00514861
CAS Number: 7078-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR