7-Hydroxy-6-methoxy-4-methyl-3-(2-morpholino-2-oxoethyl)-2-oxo-2H-chromene-8-carbaldehyde 99%

Supplier: Ambeed
Warning

A1003533-10MG A1003533-1MG A1003533-5MG A1003533-100MG A1003533-250MG
76646-168EA 99 USD
76646-168 76646-170 76646-172 77782-721 77782-722
7-Hydroxy-6-methoxy-4-methyl-3-(2-morpholino-2-oxoethyl)-2-oxo-2H-chromene-8-carbaldehyde 99%
7-Hydroxy-6-methoxy-4-methyl-3-(2-morpholino-2-oxoethyl)-2-oxo-2H-chromene-8-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₇
MDL Number: MFCD31540824
CAS Number: 1338934-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR