α-Ionone 80%

Supplier: Ambeed
Danger

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

A1003466-100G A1003466-25G A1003466-500G
76646-152EA 26.53 USD
76646-152 76646-154 76646-156
α-Ionone 80%
α-Ionone

Precaución: for Research Use Only

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
MDL Number: MFCD00001565
CAS Number: 127-41-3
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR