Ethyl toluene-4-sulfonate 90%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester

A1003456-100G A1003456-10G A1003456-25G A1003456-500G A1003456-5G
76646-142EA 11.67 USD
76646-142 76646-144 76646-146 76646-148 76646-150
Ethyl toluene-4-sulfonate 90%
Ethyl toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
MDL Number: MFCD00009100
CAS Number: 80-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR