5-Bromopyridine-3-thiol hydrochloride 98%

Supplier: Ambeed
Warning

A1003384-100MG A1003384-1G A1003384-250MG
76646-132EA 152.77 USD
76646-132 76646-134 76646-136
5-Bromopyridine-3-thiol hydrochloride 98%
5-Bromopyridine-3-thiol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₅BrClNS
MW: 226.5219 g/mol
MDL Number: MFCD34178726
CAS Number: 2665660-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR