9,9'-Diethyl-9H,9'H-3,3'-bicarbazole 97%

Supplier: Ambeed
Warning

A1003357-1G A1003357-250MG
76646-124EA 55.17 USD
76646-124 76646-126
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole 97%
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole

Precaución: for Research Use Only

Formula: C₂₈H₂₄N₂
MW: 388.51 g/mol
MDL Number: MFCD32062883
CAS Number: 20466-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR