Benzyl 1,4-dioxa-7-azaspiro[4.5]decane-7-carboxylate 96%

Supplier: Ambeed
Warning

A1003346-1G A1003346-250MG A1003346-5G
76646-118EA 309.79 USD
76646-118 76646-120 76646-122
Benzyl 1,4-dioxa-7-azaspiro[4.5]decane-7-carboxylate 96%
Benzyl 1,4-dioxa-7-azaspiro[4.5]decane-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MW: 277.3157 g/mol
MDL Number: MFCD21606396
CAS Number: 528594-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR