(S)-4,12-Bis(4-methoxyphenyl)-[2.2]-paracyclophane 97%

Supplier: Ambeed
Warning

A1003332-100MG A1003332-1G A1003332-250MG
76646-112EA 175.05 USD
76646-112 76646-114 77619-500
(S)-4,12-Bis(4-methoxyphenyl)-[2.2]-paracyclophane 97%
(S)-4,12-Bis(4-methoxyphenyl)-[2.2]-paracyclophane

Precaución: for Research Use Only

Formula: C₄₄H₄₂O₄P₂
MW: 696.75 g/mol
CAS Number: 364796-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR