Benzyl 4-propylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A1003327-100MG A1003327-250MG A1003327-5G
76646-106EA 21.22 USD
76646-106 76646-108 76646-110
Benzyl 4-propylpiperazine-1-carboxylate 97%
Benzyl 4-propylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MW: 262.3474 g/mol
CAS Number: 856844-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR