(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride, Z-Lys-OMe·HCl

A1003288-100MG A1003288-1G A1003288-250MG A1003288-5G
76646-096EA 29 USD
76646-096 76646-098 76646-100 76646-102
(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
(S)-Methyl-6-amino-2-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₂₃ClN₂O₄
MDL Number: MFCD00077025
CAS Number: 26348-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR