5-Cyclopropyl-isoxazole-3-carboxylic acid 90%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclopropylisoxazole-3-carboxylic acid

A1003286-100MG A1003286-1G A1003286-250MG A1003286-5G
76646-088EA 16.98 USD
76646-088 76646-090 76646-092 76646-094
5-Cyclopropyl-isoxazole-3-carboxylic acid 90%
5-Cyclopropyl-isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD05668325
CAS Number: 110256-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR