(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl Succinimidyl Carbonate 97%

Supplier: Ambeed
Warning

Synonyms: endo-BCN-NHS carbonate, BCN-CO-NHS, BCN-NHS, rel-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl (2,5-dioxopyrrolidin-1-yl) carbonate

A1003284-100MG A1003284-50MG A1003284-1G A1003284-250MG A1003284-5G
76646-084EA 294.94 USD
76646-084 76646-086 77548-866 77548-868 77565-742
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl Succinimidyl Carbonate 97%
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyl Succinimidyl Carbonate

Precaución: For research use only.

Formula: C₁₅H₁₇NO₅
MW: 291.30 g/mol
MDL Number: MFCD19705416
CAS Number: 1426827-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR