(S)-3-(Methoxymethyl)morpholine 98%

Supplier: Ambeed
Danger

A1003268-100MG A1003268-1G A1003268-250MG
76646-078EA 47.75 USD
76646-078 76646-080 76646-082
(S)-3-(Methoxymethyl)morpholine 98%
(S)-3-(Methoxymethyl)morpholine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.1729 g/mol
MDL Number: MFCD14586376
CAS Number: 218595-25-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR