(5-Chloro-2-methoxybenzyl)hydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A1003257-1G A1003257-250MG
76646-068EA 17 USD
76646-068 76646-070
(5-Chloro-2-methoxybenzyl)hydrazine hydrochloride 97%
(5-Chloro-2-methoxybenzyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂O
MW: 223.0997 g/mol
MDL Number: MFCD32854045
CAS Number: 2682112-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR